Skip to Content
Merck
All Photos(1)

Key Documents

M65009

Sigma-Aldrich

N-Methyloctadecylamine

98%

Synonym(s):

N-Methylstearylamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$103.00

$103.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$103.00

About This Item

Linear Formula:
CH3(CH2)17NHCH3
CAS Number:
Molecular Weight:
283.54
Beilstein:
1758431
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$103.00


0 left in stock

Request a Bulk Order

Assay

98%

mp

42-46 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCNC

InChI

1S/C19H41N/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-2/h20H,3-19H2,1-2H3

InChI key

SZEGKVHRCLBFKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service