Skip to Content
Merck
All Photos(3)

Key Documents

M39652

Sigma-Aldrich

1-Methylcyclopentanol

≥99%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$124.00
50 G
$586.00

$124.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$124.00
50 G
$586.00

About This Item

Linear Formula:
CH3C5H8OH
CAS Number:
Molecular Weight:
100.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$124.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99%

bp

135-136 °C (lit.)

mp

36-37 °C (lit.)

density

0.904 g/mL at 25 °C (lit.)

SMILES string

CC1(O)CCCC1

InChI

1S/C6H12O/c1-6(7)4-2-3-5-6/h7H,2-5H2,1H3

InChI key

CAKWRXVKWGUISE-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service