Skip to Content
Merck
All Photos(3)

Key Documents

M37935

Sigma-Aldrich

2-Methyl-1,3-cyclohexanedione

97%

Synonym(s):

1-Methyl-2,6-cyclohexanedione

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$218.00
100 G
$799.00

$218.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$218.00
100 G
$799.00

About This Item

Linear Formula:
CH3C6H7(=O)2
CAS Number:
Molecular Weight:
126.15
Beilstein:
1281598
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$218.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

206-208 °C (lit.)

SMILES string

CC1C(=O)CCCC1=O

InChI

1S/C7H10O2/c1-5-6(8)3-2-4-7(5)9/h5H,2-4H2,1H3

InChI key

VSGJHHIAMHUZKF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

An important starting material, exemplified in a formal synthesis of acorone.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The Journal of Organic Chemistry, 58, 2972-2972 (1993)
Randy M Wadkins et al.
Journal of medicinal chemistry, 48(8), 2906-2915 (2005-04-15)
Carboxylesterases (CE) are ubiquitous enzymes responsible for the metabolism of xenobiotics. Because the structural and amino acid homology among esterases of different classes, the identification of selective inhibitors of these proteins has proved problematic. Using Telik's target-related affinity profiling (TRAP)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service