Skip to Content
Merck
All Photos(1)

Key Documents

M11006

Sigma-Aldrich

3-Methoxybenzyl alcohol

98%

Synonym(s):

m-Anisyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$99.60

$99.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$99.60

About This Item

Linear Formula:
CH3OC6H4CH2OH
CAS Number:
Molecular Weight:
138.16
Beilstein:
2042063
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$99.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.540 (lit.)

bp

250 °C/723 mmHg (lit.)

density

1.112 g/mL at 25 °C (lit.)

SMILES string

COc1cccc(CO)c1

InChI

1S/C8H10O2/c1-10-8-4-2-3-7(5-8)6-9/h2-5,9H,6H2,1H3

InChI key

IIGNZLVHOZEOPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elyse A Baroncini et al.
International journal of biological macromolecules, 113, 1041-1051 (2018-03-06)
Growing environmental and economic concerns as well as the uncertainty that accompanies finite petrochemical resources contributes to the increase in research and development of bio-based, renewable polymers. Concurrently, industrial and consumer demand for smaller, safer, and more flexible technologies motivates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service