Skip to Content
Merck
All Photos(2)

Documents

L556

Sigma-Aldrich

Dodecanoic acid

98%

Synonym(s):

ABL, Lauric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10COOH
CAS Number:
Molecular Weight:
200.32
Beilstein:
1099477
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 121 °C)
50 mmHg ( 210 °C)

Assay

98%

bp

225 °C/100 mmHg (lit.)

mp

44-46 °C (lit.)

density

0.883 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCC(O)=O

InChI

1S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)

InChI key

POULHZVOKOAJMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dodecanoic acid is a saturated fatty acid used as a starting material or intermediate in the synthesis of pharmaceuticals.

Application

Dodecanoic acid has been used as a reagent to synthesize MnFe2O4 magnetic nanoparticles by seed mediated growth method. It can undergo esterification with 2-ethylhexanol in the presence of sulfated zirconia catalyst to form 2-ethylhexanoldodecanoate, a biodiesel.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

348.8 °F - closed cup

Flash Point(C)

176 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid Acid Catalysts for Biodiesel Production-Towards Sustainable Energy.
Kiss AA, et al.
advanced synthesis and catalysis, 348(1-2), 75-81 (2006)
Preparation of pyrenyl-based multifunctional nanocomposites for biomedical applications.
Lim EK and Chung BH
Nature Protocols, 11(2), 236-251 (2016)
Isaac Levy et al.
The Journal of clinical endocrinology and metabolism, 100(11), 4106-4113 (2015-09-01)
Adults with hypoparathyroidism have significant rates of nephrocalcinosis and impaired renal function. Little is known about the impact of hypoparathyroidism treatment on renal function in children. To determine the prevalence and predictors for renal abnormalities (nephrocalcinosis and decreased estimated glomerular
Esther Mesman et al.
Bipolar disorders, 17(1), 39-49 (2014-07-22)
There is increasing evidence that both immune and neurochemical alterations are involved in the pathogenesis of bipolar disorder; however, their precise role remains unclear. In this study, we aimed to evaluate neuro-immune changes in a prospective study on children of
Smritilekha Bera et al.
Journal of medicinal chemistry, 51(19), 6160-6164 (2008-09-10)
Aminoglycoside antibiotics and cationic detergents constitute two classes of clinically important drugs and antiseptics. Their bacteriological and clinical efficacy, however, has decreased recently due to antibiotic resistance. We have synthesized aminoglycoside-lipid conjugates in which the aminoglycoside neomycin forms the cationic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service