Skip to Content
Merck
All Photos(1)

Key Documents

L156884

4-TERT-BUTYL-2,6-BIS(2,6-DICHLOROBENZYLIDENE)CYCLOHEXANONE

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
USD 5.75
25 G
USD 9.20
100 G
USD 11.50
500 G
USD 25.30
1 KG
USD 46.00

Vendor SKU: A145351-5G

USD 5.75


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
5 G
USD 5.75
25 G
USD 9.20
100 G
USD 11.50
500 G
USD 25.30
1 KG
USD 46.00

About This Item

Linear Formula:
C24H22Cl4O
CAS Number:
Molecular Weight:
468.254
MDL number:

Vendor SKU: A145351-5G

USD 5.75


Ships in 7 days - from Aldrich Partner.

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service