Skip to Content
Merck
All Photos(1)

Key Documents

L050082

Sigma-Aldrich

Cap B

Sign Into View Organizational & Contract Pricing

Select a Size

45 L
$6,300.00

$6,300.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
45 L
$6,300.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.21

$6,300.00


Estimated to ship on29 May 2025


Request a Bulk Order

product line

Proligo Reagents

Quality Level

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Tetrahydrofurane / N-Methylimidazole / Pyridine 8/1/1 (v/v/v)

Other Notes

Cap B solution, containing tetrahydrofuran (THF), pyridine and N-methylimidazole (NMI), is mixed in situ with Cap A solution during the capping (acetylation) reaction. Pyridine is applied as a mild base while NMI provides a powerful acylation catalyst.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

6.1 °F

Flash Point(C)

-14.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service