Skip to Content
Merck
All Photos(1)

Key Documents

L020001-HH

Sigma-Aldrich

DCA Deblock

with Dichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.21
Pricing and availability is not currently available.

type

for DNA synthesis

product line

Proligo Reagents

packaging

pkg of 4 × 2.5 L (L020251-04)

concentration

42.2-51.6 g/L (DCA content, titration)

impurities

≤200 ppm water content (Karl Fischer)

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Dicloromethane / Dichloroacetic acid 97/3 (v/v)

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service