Skip to Content
Merck
All Photos(1)

Key Documents

JKA3037

Sigma-Aldrich

mPEG5K

average MN 5,000, Silane

Synonym(s):

mPEG-Silane, Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,140.00

$1,140.00


Available to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$1,140.00

About This Item

Linear Formula:
CH3O(CH2CH2O)nCH2CONHCH2CH2CH2Si(OCH2CH3)3
UNSPSC Code:
12162002
NACRES:
NA.23

$1,140.00


Available to ship on28 April 2025Details


Request a Bulk Order

Product Name

mPEG5K-Silane, average Mn 5,000

form

solid

mol wt

average Mn 5,000

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

General description

Methoxy PEG Silane for glass or silica surface modification and deactivation.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kobayashi, Y., et al.
ISRN Nanomaterials, 2013 (2013)

Questions

  1. Will mPeg-silane survive  after 10mM NaOH wash? 

    1 answer
    1. When Product JKA3037 is used to silanize a surface, the functional group attaching it to that surface is base-labile. So one would not expect it to survive after a wash with a 10 mM sodium hydroxide solution (approximately pH 12). Consider using a milder solution such as 100 mM sodium bicarbonate (pH approximately 8.2-8.4).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service