Skip to Content
Merck
All Photos(1)

Key Documents

JKA0001

Sigma-Aldrich

Y-PEG40K

average MN 40,000, NHS, amine reactive

Synonym(s):

Y-PEG-NHS

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$580.00

$580.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$580.00

About This Item

Linear Formula:
((CH3O(CH2CH2O)nCH2CH2)(CH3(CH2CH2O)nCH2CO)NCH2COOC4H4O2N
UNSPSC Code:
12162002
NACRES:
NA.23

$580.00


Check Cart for Availability

Request a Bulk Order

Product Name

Y-PEG40K-NHS, average Mn 40,000

form

powder

mol wt

average Mn 40,000

reaction suitability

reactivity: amine reactive

polymer architecture

shape: Y-shape
functionality: monofunctional

General description

Y-shape PEG NHS from JenKem Technology is a Succinimidyl Carboxymethyl Ester with two PEG arms, reactive towards the amino group of lysine(s) on proteins or other biologics. Amine PEGylation with Y-shape PEG NHS can be completed in less than 1hr at pH 7-8. JenKem proprietary Y-shape PEGs are more selective, due to their sterically bulky structure.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service