All Photos(1)
About This Item
Linear Formula:
ICH2CO2H
CAS Number:
Molecular Weight:
185.95
Beilstein:
1739079
EC Number:
MDL number:
UNSPSC Code:
12352200
Recommended Products
Assay
98%
mp
77-79 °C (lit.)
SMILES string
OC(=O)CI
InChI
1S/C2H3IO2/c3-1-2(4)5/h1H2,(H,4,5)
InChI key
JDNTWHVOXJZDSN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Enolic radical and ketene precursor. SH modifier.
Reagent for the modification of cysteine residues in proteins.
replaced by
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1A
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tetrahedron, 49, 8465-8465 (1993)
R Nemani et al.
Archives of biochemistry and biophysics, 300(1), 24-29 (1993-01-01)
Chemical modification of the sulfhydryl residues of the catalytic subunits of protein phosphatases 1 and 2A was studied. Both enzymes were inactivated by a variety of thiol group reagents. Mercurial compounds were the most effective inactivators. Of the alkylating agents
Tetrahedron, 50, 7579-7579 (1994)
Catherine E Ferland et al.
Phytotherapy research : PTR, 26(9), 1278-1285 (2012-09-28)
The aim of the present study was to evaluate whether eugenol, the main constituent of clove oil, has the capacity to provide analgesia in the monoiodoacetate-induced rat model of osteoarthritis. Animals (n = 6/group) received either eugenol (20 or 40 mg/kg) or a
Tatyana Y Samgina et al.
Journal of the American Society for Mass Spectrometry, 22(12), 2246-2255 (2011-10-08)
Mass spectrometry faces considerable difficulties in de novo sequencing of long non-tryptic peptides with S-S bonds. Long disulfide-containing peptides brevinins 1E and 2Ec from frog Rana ridibunda were reduced and alkylated with nine novel and three known derivatizing agents. Eight
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service