Skip to Content
Merck
All Photos(1)

Key Documents

I15807

Sigma-Aldrich

Isochroman

99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$225.00

$225.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$225.00

About This Item

Empirical Formula (Hill Notation):
C9H10O
CAS Number:
Molecular Weight:
134.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$225.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.545 (lit.)

density

1.067 g/mL at 25 °C (lit.)

SMILES string

C1Cc2ccccc2CO1

InChI

1S/C9H10O/c1-2-4-9-7-10-6-5-8(9)3-1/h1-4H,5-7H2

InChI key

HEBMCVBCEDMUOF-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin P Cole et al.
Journal of the American Chemical Society, 142(31), 13573-13581 (2020-07-15)
The Birch reduction is a powerful synthetic methodology that uses solvated electrons to convert inert arenes to 1,4-cyclohexadienes-valuable intermediates for building molecular complexity. Birch reductions traditionally employ alkali metals dissolved in ammonia to produce a solvated electron for the reduction
Shah-Iram Niaz et al.
Natural product research, 33(9), 1262-1268 (2018-05-05)
Seven polyketides, including two new isochromanes, penisochromanes A and B (1 and 2), as well as five known compounds were obtained from an ascidian-derived fungus Penicillium sp. 4829. Their structures were identified by extensive spectroscopic analyses. The structures of compounds
Samantha E Yohn et al.
Pharmacology, biochemistry, and behavior, 135, 217-226 (2015-05-30)
Mesolimbic dopamine (DA), particularly in the nucleus accumbens, is a critical component of the brain circuitry involved in behavioral activation and effort-related processes. Although much is known about the characteristics of DA D2 receptor antagonism on effort-related choice behavior, less

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service