Skip to Content
Merck
All Photos(1)

Key Documents

HYASKIT

Sigma-Aldrich

HyasKit

Synonym(s):

Microenvironment mapping technology, Photoinduced proximity labeling kit, Protein-protein interaction elucidation technology, Small molecule-photocatalyst conjugate labeling

Sign Into View Organizational & Contract Pricing

Select a Size

HYASKIT-A
$9,840.00

$9,840.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
HYASKIT-A
$9,840.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.32

$9,840.00


Check Cart for Availability

Request a Bulk Order

storage temp.

−20°C

General description

The Hyas MicroMap Kit provides a method for small molecule intracellular target identification via photocatalytic proximity labeling. The kit contains a catalyst that can be coupled to an amine on almost any small-molecule ligand. The subsequent ligand-Iridium conjugate can be used to perform multiple target identification experiments. The following kit provides enough Ir-catalyst to synthesize 20 μMoles of iridium–ligand conjugate and enough Diazirine-PEG3-Biotin to perform 10 proteomics experiments.
The kit provides a step-by-step procedure for conjugating your ligand of choice to a photocatalyst that localizes light-mediated crosslinking to the ligands target. The ligand–iridium conjugate can then be incubated with live cells along with a biotinylated diazirine reagent. Analogous to a satellite, the photocatalyst localizes carbene generation from the diazirine only in the direct vicinity and thus covalently modifies the target protein in a catalytic manner, amplifying the signal for downstream analysis.
To link to an acid on a small molecule (instead of an amine), the Dione Kit is available.

Photoreactors commercially available:

Application

Proximity protein labeling, protein-protein interactions, photoaffinity labeling, proteomics

Biochem/physiol Actions

Kit of components packaged in individual vials for enabling photoinduced proximity labeling. The kit contains an Ir-catalyst to couple to an acid on a small molecule ligand. Kit contains Ir-catalyst and Diazirine PEG3 Biotin to perform 10 proteomics experiments. To link to an acid on a small molecule, the Dione Kit is available.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In cases where photoaffinity labeling (PAL) probes are ineffective, Hyas and Dione Kits provide an effective method for small molecule identification by using an Iridium photocatalyst tethered to the molecule under investigation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service