Skip to Content
Merck
All Photos(3)

Key Documents

H24402

Sigma-Aldrich

2-Hydroxycyclohexanone dimer

97%

Synonym(s):

Adipoin, NSC 117392

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$109.00
25 G
$298.00

$109.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$109.00
25 G
$298.00

About This Item

Linear Formula:
[HOC6H9(=O)]2
CAS Number:
Molecular Weight:
228.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$109.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

bp

83-86 °C/13 mmHg (lit.)

mp

100-107 °C (lit.)

storage temp.

2-8°C

SMILES string

OC12CCCCC1OC3(O)CCCCC3O2

InChI

1S/C12H20O4/c13-11-7-3-1-5-9(11)15-12(14)8-4-2-6-10(12)16-11/h9-10,13-14H,1-8H2

InChI key

OLLWHBLCQGPTMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Synthesis of substrated for tin tetrachloride mediated fragmentation to produce ynones and ynoates
  • Tandem oxidation coupling reactions with diamines
  • Asymmetric allylic alkylation of enolates
  • Formate reduction of allylic carbonates for polypropionate systems

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service