Skip to Content
Merck
All Photos(2)

Key Documents

GF82587426

Tantalum

tube, 1000mm, outside diameter 2.0mm, inside diameter 1mm, wall thickness 0.5mm, as drawn, 99.9%

Synonym(s):

Tantalum, TA007100, Ta

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$4,270.00

$4,270.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$4,270.00

About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

$4,270.00


Estimated to ship on21 May 2025


Request a Bulk Order

vapor pressure

<0.01 mmHg ( 537.2 °C)

Assay

99.9%

form

tubes

autoignition temp.

572 °F

manufacturer/tradename

Goodfellow 825-874-26

resistivity

13.5 μΩ-cm, 20°C

L × wall thickness

1000 mm × 0.5 mm

O.D. × I.D.

2.0 mm × 1.0 mm

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Black
Clinical materials, 16(3), 167-173 (1993-12-09)
A detailed literature search was carried out to define the current knowledge about the biological performance of tantalum. The pure metal appears, to a great degree, to be inert both in vivo and in vitro. Both the pure metal and
Robert Cohen
American journal of orthopedics (Belle Mead, N.J.), 31(4), 216-217 (2002-05-15)
Conventional materials used for orthopedic reconstruction implants limit the design of new and improved implants. Solid metal is rigid. Porous fixation surfaces have low volumetric porosity available for biologic ingrowth and low frictional characteristics for initial implant stability and have
Brett Levine et al.
The journal of knee surgery, 20(3), 185-194 (2007-08-02)
Porous tantalum represents an alternative metal for primary and revision total knee arthroplasty (TKA) with several unique properties. Tantalum is a transition metal, which in its bulk form has shown excellent biocompatibility and is safe to use in vivo as
Nilesh Patil et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 89(1), 242-251 (2008-10-08)
Conventional porous-coated joint prostheses used in hip and knee reconstruction have demonstrated good clinical results, however, these implants possess some inherent shortcomings such as low volumetric porosity, suboptimal frictional characteristics, and higher modulus of elasticity relative to that of bone.
Paul S Issack
The Journal of bone and joint surgery. American volume, 95(21), 1981-1987 (2013-11-08)
➤ Over the past decade, porous tantalum has emerged as a powerful tool for reconstruction of the failed acetabular component.➤ The increased porosity, high coefficient of friction, and favorable elastic modulus of porous tantalum compared with traditional titanium mesh or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service