Skip to Content
Merck
All Photos(2)

Key Documents

GF79742953

Nickel

wire reel, 1m, diameter 0.05mm, as drawn, 99.99+%

Synonym(s):

Nickel, NI005115

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$408.00

$408.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$408.00

About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23

$408.00


Check Cart for Availability

Request a Bulk Order

Assay

99.99%

form

wire

manufacturer/tradename

Goodfellow 797-429-53

resistivity

6.97 μΩ-cm, 20°C

L × diam.

1 m × 0.05 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal Cox et al.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics, 145(4), 505-513 (2014-04-08)
Nickel-titanium closed-coil springs are purported to deliver constant forces over extended ranges of activation and working times. In-vivo studies supporting this claim are limited. The objective of this study was to evaluate changes in force-decay properties of nickel-titanium closed-coil springs
Egor Geist et al.
Natural product reports, 31(4), 441-448 (2014-02-28)
This Highlight covers the current status of relatively unexplored sp(3)-sp(3) cross-coupling reactions with particular focus on natural product and related syntheses.
Giovanna Visioli et al.
Bulletin of environmental contamination and toxicology, 92(4), 490-496 (2013-11-30)
In vitro short-term chronic phytotoxicity germination and root elongation test were applied to test the effects of nickel (Ni) in seed germination and root elongation in six plants species: Cucumis sativus (Cucurbitaceae), Lepidium sativum and Brassica nigra (Brassicaceae), Trifolium alexandrinum
Gonensin O Bozdag et al.
Gene, 538(2), 251-257 (2014-02-04)
Nickel is an essential micronutrient due to its involvement in many enzymatic reactions as a cofactor. However, excess of this element is toxic to biological systems. Here, we constructed a cDNA library from Beta maritima and screened it in the
Daniel H Rothman et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(15), 5462-5467 (2014-04-08)
The end-Permian extinction is associated with a mysterious disruption to Earth's carbon cycle. Here we identify causal mechanisms via three observations. First, we show that geochemical signals indicate superexponential growth of the marine inorganic carbon reservoir, coincident with the extinction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service