Skip to Content
Merck
All Photos(1)

Key Documents

GF70980813

Polymethylmethacrylate

(PMMA, ACRYLIC), powder, mean particle size 48micron, weight 500 g, molecular weight 900-1500k g/mol

Synonym(s):

ME306048

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$593.00

$593.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$593.00

About This Item

UNSPSC Code:
12352100
NACRES:
NA.23

$593.00


Check Cart for Availability

Request a Bulk Order

form

powder

manufacturer/tradename

Goodfellow 709-808-13

weight

500 g

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Just to confirm, is the molecular weight range 900k to 1500k or 900 to 1500k grams per mole? Thanks

    1 answer
    1. The molecular weight range for this product is 900-1500k g/mol.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service