Skip to Content
Merck
All Photos(2)

Key Documents

GF70640838

Beryllium

foil, thickness 0.00025 mm, size 25 × 25 mm, purity 99.9%

Synonym(s):

BE000050, Glucinium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Be
CAS Number:
Molecular Weight:
9.01
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

foil

autoignition temp.

1198 °F

manufacturer/tradename

Goodfellow 706-408-38

resistivity

4.46 μΩ-cm, 20°C

size

25 × 25 mm

thickness

0.00025 mm

bp

2970 °C (lit.)

mp

1278 °C (lit.)

density

1.85 g/mL at 25 °C (lit.)

SMILES string

[Be]

InChI

1S/Be

InChI key

ATBAMAFKBVZNFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service