Skip to Content
Merck
All Photos(1)

Key Documents

GF70145984

Thoriated tungsten

wire, 0.15 mm diameter, W 99%/Th 1%, length 25 m, temper as drawn

Synonym(s):

W 145351

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥99%

form

wire

manufacturer/tradename

Goodfellow 701-459-84

L

25 m

diameter

0.15 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Does it come with the tungsten carbide layer already on the surface?

    1 answer
    1. This material has been etched to a clean condition and therefore will not have a tungsten carbide outer layer.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service