GF68848521
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3
mesh, 150 x 150mm, 0.103mm nominal aperture, 0.066mm wire diameter, 150 x 150 wires/inch, 0.37 open area, plain weave mesh type
Synonym(s):
FE248705
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
150 x 150 (wires/inch)
form
(mesh)
manufacturer/tradename
Goodfellow 688-485-21
diam. × nominal capacity
0.066 mm × 0.103 mm , plain weave mesh
size × opening
150 mm × 150 mm × 0.37
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service