GF61256022
Nimonic® alloy 90, Ni53Cr20Co18Ti2.5Al1.5Fe1.5
rod, 500mm x 22mm diameter
Synonym(s):
NJ107950
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101712
NACRES:
NA.23
form
rod
manufacturer/tradename
Goodfellow 612-560-22
L × diam.
500 mm × 22 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
Nimonic is a registered trademark of Special Metals Corporation
Nimonic is a registered trademark of Special Metals Corporation
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service