Skip to Content
Merck
All Photos(2)

Key Documents

GF48095185

Dysprosium

foil, 50x50mm, thickness 0.125mm, as rolled, 99%

Synonym(s):

Dysprosium, DY000225

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,780.00

$1,780.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$1,780.00

About This Item

Empirical Formula (Hill Notation):
Dy
CAS Number:
Molecular Weight:
162.50
MDL number:
UNSPSC Code:
12141602
PubChem Substance ID:
NACRES:
NA.23

$1,780.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

foil

manufacturer/tradename

Goodfellow 480-951-85

resistivity

89 μΩ-cm, 20°C

size × thickness

50 x 50 mm × 0.125 mm

bp

2567 °C (lit.)

mp

1412 °C (lit.)

density

8.559 g/mL at 25 °C (lit.)

SMILES string

[Dy]

InChI

1S/Dy

InChI key

KBQHZAAAGSGFKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chelates of gadolinium and dysprosium as contrast agents for MR imaging.
S M Rocklage et al.
Journal of magnetic resonance imaging : JMRI, 3(1), 167-178 (1993-01-01)
MRI contrast agents based on dysprosium or holmium.
Małgorzata Norek et al.
Progress in nuclear magnetic resonance spectroscopy, 59(1), 64-82 (2011-05-24)
Sabine Tricot et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 87(4), 357-368 (2015-02-24)
The recent introduction of mass cytometry, a technique coupling a cell introduction system generating a stream of single cells with mass spectrometry, has greatly increased the number of parameters that can be measured per single cell. As with all new
Daiki Goto et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 31(11), 1143-1149 (2015-11-13)
It is difficult to control the electrophoretic mobility in order to obtain high resolution among saccharides in complex samples. We report herein on a new affinity capillary electrophoresis (ACE) method for an anionic monosaccharide, N-acetylneuraminic acid (Neu5Ac), which is important
Bieke Onghena et al.
Dalton transactions (Cambridge, England : 2003), 43(30), 11566-11578 (2014-06-19)
The ionic liquid choline bis(trifluoromethylsulfonyl)imide, [Chol][Tf2N], was used for the extraction of neodymium(III), in combination with choline hexafluoroacetylacetonate, [Chol][hfac], as the extractant. The binary mixture of [Chol][Tf2N] and water shows temperature-dependent phase behavior, with an upper critical solution temperature of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service