Skip to Content
Merck
All Photos(2)

Key Documents

GF45482283

Zinc

foil, 1m coil, thickness 1.0mm, as rolled, 99.95+%

Synonym(s):

Zinc, ZN000322

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,180.00

$1,180.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$1,180.00

About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23

$1,180.00


Check Cart for Availability

Request a Bulk Order

vapor pressure

1 mmHg ( 487 °C)

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 454-822-83

resistivity

5.8 μΩ-cm, 20°C

L × thickness

1 m × 1.0 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new vapor-phase hydrothermal method to concurrently grow ZnO nanotube and nanorod array films on different sides of a zinc foil substrate.
Haimin Zhang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(17), 5165-5169 (2012-03-21)
Xufeng Wu et al.
Chemical communications (Cambridge, England), (15)(15), 1655-1657 (2006-04-04)
Highly oriented ZnO nanoneedle/nanorods arrays have been fabricated by direct oxidation of zinc foil in alkaline zincate ion solution at near room temperature (20 degrees C for nanoneedles, 30 degrees C for nanorods).
Chenglin Yan et al.
The journal of physical chemistry. B, 110(51), 25850-25855 (2006-12-22)
Vertically aligned ZnO nanorods with uniform diameter and length have been synthesized on a zinc foil substrate with ammonium persulfate as oxidant via a facile, larger scale production and inexpensively synthesized method without any templates or additives. SEM and XRD
Chuanhui Xia et al.
Journal of nanoscience and nanotechnology, 11(12), 10506-10510 (2012-03-14)
We have investigated the properties of Mn-doped ZnO nanocrystalline film growing on zinc foil by the hydrothermal method. X-ray photoelectron spectroscopy shows that the manganese ions exist as Mn2+ in the film. From UV-vis spectra, we observe a red shift
Tandra Ghoshal et al.
Nanotechnology, 19(6), 065606-065606 (2008-02-13)
ZnO nanowire arrays were synthesized on zinc foil by a simple thermal evaporation process at relatively low temperature. Morphology and size controlled synthesis of the ZnO nanostructures was achieved by variation of the synthesis temperature, reaction time and the surface

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service