Skip to Content
Merck
All Photos(2)

Key Documents

GF38000281

Zirconium

foil, 2m coil, thickness 0.025mm, annealed, 99.2%

Synonym(s):

Zirconium, ZR000210

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$3,110.00

$3,110.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$3,110.00

About This Item

Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22
MDL number:
UNSPSC Code:
12141751
PubChem Substance ID:
NACRES:
NA.23

$3,110.00


Check Cart for Availability

Request a Bulk Order

Assay

99.2%

form

foil

manufacturer/tradename

Goodfellow 380-002-81

resistivity

40 μΩ-cm, 20°C

L × thickness

2 m × 0.025 mm

bp

4377 °C (lit.)

mp

1852 °C (lit.)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[Zr]

InChI

1S/Zr

InChI key

QCWXUUIWCKQGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Velázquez-Cayón et al.
Medicina oral, patologia oral y cirugia bucal, 17(2), e246-e250 (2011-12-07)
The increase of aesthetic demands, together with the successful outcome of current implants, has renewed interest in the search for new materials with enough mechanical properties and better aesthetic qualities than the materials customarily used in implanto-prosthetic rehabilitation. Among these
Glypican-3-targeted 89Zr PET imaging of hepatocellular carcinoma: where antibody imaging dares to tread.
Bernadette V Marquez et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(5), 708-709 (2014-03-26)
Anjana Raut et al.
Indian journal of dental research : official publication of Indian Society for Dental Research, 22(1), 140-143 (2011-04-29)
The demand for esthetic restorations has resulted in an increased use of dental ceramics for anterior and posterior restorations. A few decades ago, all-ceramic restorations were restricted to treatment in the anterior region, but now all-ceramic restorations can be made
Yin Zhang et al.
Current radiopharmaceuticals, 4(2), 131-139 (2011-12-24)
Positron emission tomography (PET) imaging with radiolabeled monoclonal antibodies has always been a dynamic area in molecular imaging. With decay half-life (3.3 d) well matched to the circulation half-lives of antibodies (usually on the order of days), (89)Zr has been
Joseph R Osborne et al.
The Journal of urology, 191(5), 1439-1445 (2013-10-19)
In this pilot study we explored the feasibility of (89)Zr labeled J591 monoclonal antibody positron emission tomography of localized prostate cancer. Before scheduled radical prostatectomy 11 patients were injected intravenously with (89)Zr-J591, followed 6 days later by whole body positron

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service