Skip to Content
Merck
All Photos(2)

Key Documents

GF37754859

Platinum

foil, 0.05m coil, thickness 0.2mm, coil width 5mm, as rolled, 99.95%

Synonym(s):

Platinum, PT000265

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,450.00

$1,450.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$1,450.00

About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23

$1,450.00


Check Cart for Availability

Request a Bulk Order

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 377-548-59

resistivity

10.6 μΩ-cm, 20°C

L × thickness × width

0.05 m × 0.2 mm × 5 mm

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily L Foldes et al.
Journal of neuroscience methods, 196(1), 31-37 (2010-12-29)
Nerve cuff electrodes are a principle tool of basic and applied electro-neurophysiology studies and are championed for their ability to achieve good nerve recruitment with low thresholds. We describe the design and method of fabrication for a novel circumpolar peripheral
S M Hannon et al.
The International journal of prosthodontics, 4(5), 457-464 (1991-09-01)
Postceramic soldering of collarless veneered retainers in which the porcelain margins were formed with a platinum foil matrix technique was investigated. The purpose of the study was to determine if removing the platinum foil matrix before or after indexing and
L Tröger et al.
Journal of synchrotron radiation, 8(Pt 2), 105-109 (2001-08-22)
Experimental data are presented which demonstrate the existence of a fine structure in extended X-ray absorption spectra due to interference effects in the initial photon state (piXAFS). Interference occurs between the incident electromagnetic wave and its coherently scattered waves from
Sameer Puri
Compendium of continuing education in dentistry (Jamesburg, N.J. : 1995), 26(7), 519-525 (2005-08-03)
Porcelain is an increasingly popular material to use for restorations. This article will discuss the 3 main ways to fabricate porcelain restorations. The first method involves waxing up the restoration to the proper form and casting it in molten porcelain
J A Sorensen et al.
The Journal of prosthetic dentistry, 67(1), 16-22 (1992-01-01)
This study evaluated the marginal fidelity and microleakage of porcelain veneers made with the platinum foil and refractory die techniques. Maxillary incisors, matched for size and amount of enamel, were prepared with 0.5 mm uniform intraenamel reduction. The indirectly made

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service