Skip to Content
Merck
All Photos(2)

Key Documents

GF29522101

Platinum

foil, 10mm disks, thickness 0.01mm, as rolled, 99.95%

Synonym(s):

Platinum, PT000215

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 295-221-01

resistivity

10.6 μΩ-cm, 20°C

diam. × thickness

10 mm × 0.01 mm

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily L Foldes et al.
Journal of neuroscience methods, 196(1), 31-37 (2010-12-29)
Nerve cuff electrodes are a principle tool of basic and applied electro-neurophysiology studies and are championed for their ability to achieve good nerve recruitment with low thresholds. We describe the design and method of fabrication for a novel circumpolar peripheral
L Tröger et al.
Journal of synchrotron radiation, 8(Pt 2), 105-109 (2001-08-22)
Experimental data are presented which demonstrate the existence of a fine structure in extended X-ray absorption spectra due to interference effects in the initial photon state (piXAFS). Interference occurs between the incident electromagnetic wave and its coherently scattered waves from
Sameer Puri
Compendium of continuing education in dentistry (Jamesburg, N.J. : 1995), 26(7), 519-525 (2005-08-03)
Porcelain is an increasingly popular material to use for restorations. This article will discuss the 3 main ways to fabricate porcelain restorations. The first method involves waxing up the restoration to the proper form and casting it in molten porcelain
S M Hannon et al.
The International journal of prosthodontics, 4(5), 457-464 (1991-09-01)
Postceramic soldering of collarless veneered retainers in which the porcelain margins were formed with a platinum foil matrix technique was investigated. The purpose of the study was to determine if removing the platinum foil matrix before or after indexing and
Ayumi Ikeda et al.
Journal of medical and dental sciences, 52(4), 195-202 (2006-05-04)
The purpose of this study was to clarify the influence of changes of occlusal contacts on adjacent tooth displacement during articulation. The amount and direction of displacement of the maxillary left first molar and adjacent teeth were measured using a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service