Skip to Content
Merck
All Photos(1)

Key Documents

GF27430017

Polystyrene

(PS), powder, mean particle size 250micron, weight 100 g, condition homopolymer

Synonym(s):

ST316051

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100

form

powder

manufacturer/tradename

Goodfellow 274-300-17

weight

100 g

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service