Skip to Content
Merck
All Photos(1)

Key Documents

GF27051296

Arsenic

lump, 20 mm max. lump size, weight 100 g, high purity 99.9999%

Synonym(s):

AS006105

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,730.00

$1,730.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$1,730.00

About This Item

UNSPSC Code:
12141906
NACRES:
NA.23

$1,730.00


Check Cart for Availability

Request a Bulk Order

Assay

99.9999%

form

lumps

manufacturer/tradename

Goodfellow 270-512-96

weight

100 g

max. lump size

20 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Repr. 1A - STOT RE 1 Oral

Target Organs

Respiratory organs,Cardio-vascular system,Gastro-intestinal system,Central nervous system,Nervous system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service