Skip to Content
Merck
All Photos(1)

Key Documents

GF24554864

Glass Sealing Alloy, Fe54Ni29Co17

200mm tube, 3.175mm outside diameter, 0.254mm wall thickness, 2.667mm inside diameter, as drawn

Synonym(s):

FE047410

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23
Pricing and availability is not currently available.

form

tubes

manufacturer/tradename

Goodfellow 245-548-64

L × wall thickness

200 mm × 0.254 mm

O.D. × I.D.

3.175 mm × 2.667 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service