GF17441502
Stainless Steel - AISI 304 alloy, FeCr18Ni10
500mm tube, 9.0mm outside diameter, 0.5mm wall thickness, 8mm inside diameter, hard
Synonym(s):
FE227710
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
tubes
manufacturer/tradename
Goodfellow 174-415-02
L × thickness
500 mm × 0.5 mm
O.D. × I.D.
9.0 mm × 8.0 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service