Skip to Content
Merck
All Photos(1)

Key Documents

GF12567786

Nickel-iron alloy, Ni70Fe30

100m insulated wire, 0.025mm conductor diameter, 0.0035mm insulation thickness, polyimide insulation

Synonym(s):

NJ125825

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,040.00

$1,040.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$1,040.00

About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

$1,040.00


Check Cart for Availability

Request a Bulk Order

description

polyimide insulation

form

wire (insulated)

manufacturer/tradename

Goodfellow 125-677-86

L

100 m

conductor diam.

0.025 mm

insulation thickness

0.0035 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service