GF04970202
Aluminum
foil, thickness 0.125 mm, size 300 × 300 mm, purity 99%
Synonym(s):
AL000592
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Assay
99%
form
foil
autoignition temp.
1400 °F
manufacturer/tradename
Goodfellow 049-702-02
resistivity
2.6548 μΩ-cm
size
300 × 300 mm
thickness
0.125 mm
bp
2460 °C (lit.)
mp
660.37 °C (lit.)
density
2.7 g/mL at 25 °C (lit.)
SMILES string
[Al]
InChI
1S/Al
InChI key
XAGFODPZIPBFFR-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service