Skip to Content
Merck
All Photos(1)

Key Documents

GEN-7032

Sigma-Aldrich

Cationic Liposomes for DNA/RNA delivery

DC-Chol:DOPE (1:2) containing 0.5% Rhod-PE (Fluorescent)

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$1,080.00

$1,080.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$1,080.00

About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

$1,080.00


Estimated to ship on23 May 2025


Request a Bulk Order

Quality Level

contains

Rhod-PE (1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(lissamine
rhodamine B sulfonyl)) as fluorescent label

composition

Deionized RNAse-free Water

concentration

0.02 mM (Rhod-PE)
2 mM (DC-Cholesterol)
3.98 mM (DOPE)

impurities

1:2 mol/mol DC-Chol:DOPE

particle size

100 nm

pH

7

absorbance ratio

560/583 nm

Looking for similar products? Visit Product Comparison Guide

General description

Cationic liposomes are traditionally used for the delivery of genetic materials such as various types of DNA (pDNA, cDNA, CpG DNA, oligonucleotide, antisense oligonucleotide), various types of RNA such as (siRNA, mRNA) and nucleic acid mimics (NAMs). The positive, cationic charge of the lipids is used to neutralize the negative charge of nucleic acids and results in greater encapsulation efficiency, cellular uptake and endosomal delivery.

Application

Drug delivery
Gene delivery
Lipid-protein interactions

Storage and Stability

Liposomes should never be frozen. Liposomes should be stored in the dark at 4°C, except when brought to room temperature for brief periods prior to use.

Liposomes are made under sterile conditions. If you need to take multiple aliquots out of the vial, it is advised to take extreme care in not contaminating the vial. It is recommended to handle the vial under a sterile hood to maintain the sterility of the product. Liposomes should never be frozen. Ice crystals that form during freezing will rupture the lipid membrane of the liposomes and change the size of liposomes particles.

Legal Information

Genesome is a trademark of Encapsula NanoSciences
Product of Encapsula Nanosciences

Disclaimer

For research use only

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service