Skip to Content
Merck
All Photos(2)

Key Documents

F8004

Sigma-Aldrich

4-Fluorobenzyl chloride

99%

Synonym(s):

α-Chloro-4-fluorotoluene, 1-Chloromethyl-4-fluorobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$61.00
100 G
$99.90

$61.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$61.00
100 G
$99.90

About This Item

Linear Formula:
FC6H4CH2Cl
CAS Number:
Molecular Weight:
144.57
Beilstein:
742272
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$61.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.513 (lit.)

bp

82 °C/26 mmHg (lit.)

density

1.207 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(CCl)cc1

InChI

1S/C7H6ClF/c8-5-6-1-3-7(9)4-2-6/h1-4H,5H2

InChI key

IZXWCDITFDNEBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esakkiammal Sudha Esakkimuthu et al.
Molecules (Basel, Switzerland), 25(14) (2020-07-18)
Lignin is an aromatic biopolymer derived from lignocellulosic biomass. Providing a comprehensive structural analysis of lignin is the primary motivation for the quantification of various functional groups, with a view to valorizing lignin in a wide range of applications. This
J Abraham Domínguez-Avila et al.
Journal of the science of food and agriculture, 99(4), 1604-1612 (2018-09-07)
Serum lipoproteins are in dynamic equilibrium, partially controlled by the apolipoprotein A1 to apolipoprotein B ratio (APOA1/APOB). Freeze-dried mango pulp (FDM) is a rich source of phenolic compounds (MP) and dietary fiber (MF), although their effects on lipoprotein metabolism have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service