Skip to Content
Merck
All Photos(2)

Key Documents

F7202

Sigma-Aldrich

4-Fluorobenzophenone

97%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$133.00

$133.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$133.00

About This Item

Linear Formula:
FC6H4COC6H5
CAS Number:
Molecular Weight:
200.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$133.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

47-49 °C (lit.)

SMILES string

Fc1ccc(cc1)C(=O)c2ccccc2

InChI

1S/C13H9FO/c14-12-8-6-11(7-9-12)13(15)10-4-2-1-3-5-10/h1-9H

InChI key

OGTSHGYHILFRHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongkun Yu et al.
Chemistry, an Asian journal, 14(19), 3350-3356 (2019-08-27)
Eutectic molecular liquids (EMLs) based on hydrogen-bonding interaction and π-π stacking were prepared. We found that the thermodynamic properties like initial decomposition temperature and glass transition temperature of EMLs are mainly dominated by the hydrogen bond donor, which is beneficial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service