Skip to Content
Merck
All Photos(1)

Documents

F7008

Sigma-Aldrich

2-Fluorobenzophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9FO
CAS Number:
Molecular Weight:
200.21
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

98%

SMILES string

Fc1ccccc1C(=O)c2ccccc2

InChI

1S/C13H9FO/c14-12-9-5-4-8-11(12)13(15)10-6-2-1-3-7-10/h1-9H

InChI key

DWFDQVMFSLLMPE-UHFFFAOYSA-N


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service