Skip to Content
Merck
All Photos(1)

Key Documents

F20602

Sigma-Aldrich

trans-3-(2-Furyl)acrolein

97%

Synonym(s):

2-Furanacrolein

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$167.00

$167.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
25 G
$167.00

About This Item

Empirical Formula (Hill Notation):
C7H6O2
CAS Number:
Molecular Weight:
122.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$167.00


0 left in stock

Request a Bulk Order

Quality Level

Assay

97%

bp

143 °C/37 mmHg (lit.)

mp

48-50 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C\C=C\c1ccco1

InChI

1S/C7H6O2/c8-5-1-3-7-4-2-6-9-7/h1-6H/b3-1+

InChI key

VZIRCHXYMBFNFD-HNQUOIGGSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service