Skip to Content
Merck
All Photos(2)

Documents

E4641

Sigma-Aldrich

N4-Ethyl-N4-(2-hydroxyethyl)-2-methyl-1,4-phenylenediamine sulfate salt

suitable for photographic applications

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20N2O5S
CAS Number:
Molecular Weight:
292.35
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

suitability

suitable for photographic applications

SMILES string

OS(O)(=O)=O.CCN(CCO)c1ccc(N)c(C)c1

InChI

1S/C11H18N2O.H2O4S/c1-3-13(6-7-14)10-4-5-11(12)9(2)8-10;1-5(2,3)4/h4-5,8,14H,3,6-7,12H2,1-2H3;(H2,1,2,3,4)

InChI key

GVEYRUKUJCHJSR-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contact dermatitis to CD4.
F Menezes Brandão et al.
Contact dermatitis, 12(1), 48-48 (1985-01-01)
J H Liu
Zhonghua zheng xing shao shang wai ke za zhi = Zhonghua zheng xing shao shang waikf [i.e. waike] zazhi = Chinese journal of plastic surgery and burns, 6(1), 40-41 (1990-03-01)
Eleven patients burned, with CD-4-a colon darlopel were cured in our hospital from July 1978 to March 1986. Five of them were combined with acute poisoning. CD-4 is a poisonous, corrosive and irritant compound of ammonic benzene. It can be
J Schoel et al.
Der Hautarzt; Zeitschrift fur Dermatologie, Venerologie, und verwandte Gebiete, 42(4), 251-253 (1991-04-01)
We report on a single case of a lichen planus like contact dermatitis caused by colour developers. We emphasize similarities to lichen planus with regard to clinical, histopathological and immunohistological aspects.
Allergic contact dermatitis from colour developers: absence of cross-sensitivity to para-amino compounds.
P A Galindo et al.
Contact dermatitis, 30(5), 301-301 (1994-05-01)
C Lidén et al.
Contact dermatitis, 19(4), 290-295 (1988-10-01)
Colour developing agents, derivatives of p-phenylenediamine, can cause contact allergy. Patch test reactions to more than one colour developer are sometimes seen in patients. To study whether this is due to simultaneous sensitization or cross-reactivity, guinea pig maximization tests (GPMT)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service