Skip to Content
Merck
All Photos(1)

Documents

E41206

Sigma-Aldrich

Ethyl 4-nitrobenzoylacetate

Synonym(s):

(p-Nitrobenzoyl)acetic acid ethyl ester, Ethyl 3-oxo-3-(4-nitrophenyl)propanoate, NSC 62134

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4COCH2CO2C2H5
CAS Number:
Molecular Weight:
237.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

mp

71-73 °C (lit.)

SMILES string

CCOC(=O)CC(=O)c1ccc(cc1)[N+]([O-])=O

InChI

1S/C11H11NO5/c1-2-17-11(14)7-10(13)8-3-5-9(6-4-8)12(15)16/h3-6H,2,7H2,1H3

InChI key

NGRXSVFCLHVGKU-UHFFFAOYSA-N

Application

Reactant involved in:
  • Synthesis of β-1,3-dicarbonyl aldehydes via oxidation
  • Knoevenagel condensation using a lysine catalyst
  • Cycloisomerization fo rthe synthesis of trisubstituted furans
  • Fluorination by HF and iodosylbenzene
  • Intramolecular Michael addition reactions for synthesis of benzylbutyrolactones
  • Synthesis of diols via reduction of aromatic and aliphatic ketos esters
  • Cross-coupling reactions for stereoselective synthesis of unsymmetrical 1,4-enediones

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service