Skip to Content
Merck
All Photos(1)

Key Documents

E39600

Sigma-Aldrich

Ethyl myristate

97%

Synonym(s):

Ethyl tetradecanoate, Myristic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12COOC2H5
CAS Number:
Molecular Weight:
256.42
Beilstein:
1776382
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.436 (lit.)

bp

178-180 °C/12 mmHg (lit.)

mp

11-12 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCC(=O)OCC

InChI

1S/C16H32O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16(17)18-4-2/h3-15H2,1-2H3

InChI key

MMKRHZKQPFCLLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Woo-kyoung Lee et al.
Journal of controlled release : official journal of the Controlled Release Society, 84(3), 115-123 (2002-12-07)
An oil-in-water solvent evaporation method was used to prepare cyclosporin A (CyA)-loaded particles varying in size (nanoparticles, 'small-sized' microparticles, 'large-sized' microparticles), polymer compositions [poly(D,L-lactide-co-glycolic acid) (PLGA) 50/50, PLGA 85/15, poly(D,L-lactic acid) (PLA)] and additive fatty acid ester (ethyl myristate; EM).
Sven Hartwig et al.
Forensic science international, 131(2-3), 90-97 (2003-02-19)
Fatty acid ethyl esters (FAEE) can be used as alcohol markers in hair. It was investigated in this study whether this diagnostic method is disturbed by hair care and hair cosmetics. Traces of ethyl myristate, ethyl palmitate, ethyl oleate and
E Akaho et al.
Journal of pharmaceutical sciences, 70(11), 1225-1228 (1981-11-01)
Interactions of phenolic compounds 4-hexylresorcinol and 3,4-dimethylphenol with esters were studied using hexane-ester cosolvent systems by both phase solubility and partitioning methods. The data obtained by the phase solubility method were variable and could not be analyzed by any mathematical
Nadia De Giovanni et al.
Journal of analytical toxicology, 32(2), 156-159 (2008-03-13)
The diagnosis of alcoholism is a topical subject of discussion; in fact, many studies have been published on the determination of biochemical markers useful to this target. Fatty acid ethyl esters (FAEE) are minor metabolites of ethanol, and their usefulness
Neonatal hair analysis as a biomarker for in utero alcohol exposure.
Julia Klein et al.
The New England journal of medicine, 347(25), 2086-2086 (2002-12-20)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service