Skip to Content
Merck
All Photos(1)

Key Documents

E17852

Sigma-Aldrich

Ethyl 3-chloropropionate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2COOC2H5
CAS Number:
Molecular Weight:
136.58
Beilstein:
385698
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form:
liquid
Assay:
98%
Pricing and availability is not currently available. Contact Technical Service

Assay

98%

form

liquid

refractive index

n20/D 1.425 (lit.)

bp

162-163 °C (lit.)

density

1.003 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CCCl

InChI

1S/C5H9ClO2/c1-2-8-5(7)3-4-6/h2-4H2,1H3

InChI key

ZCLGVXACCAZJOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

121.1 °F

Flash Point(C)

49.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service