Skip to Content
Merck
All Photos(1)

Key Documents

DWKWGW601PMLME

Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting

label, Distilled water (in English, French, Arabic), capacity 500 mL

Synonym(s):

WGW601PMLME

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$91.40

$91.40


Estimated to ship on27 April 2025



Select a Size

Change View
5 EA
$91.40

About This Item

UNSPSC Code:
41103209
NACRES:
NB.43

$91.40


Estimated to ship on27 April 2025


feature

label Distilled water (in English, French, Arabic)

packaging

pkg of 5 ea

manufacturer/tradename

Azlon WGW601PMLME

capacity

500 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for distilled water dispensing within the laboratory

Features and Benefits

Multilingual with solvent labels including all new globally harmonised symbols, NFPA diamond, risk phrases and safety information

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service