Skip to Content
Merck
All Photos(1)

Key Documents

DWKWGW230P

Azlon Square Shoulder Unvented Wash Bottles

wide-neck, low-density polyethylene bottle, white polypropylene closure, capacity 250 mL

Synonym(s):

WGW230P

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$54.70

$54.70


Estimated to ship on24 April 2025



Select a Size

Change View
5 EA
$54.70

About This Item

UNSPSC Code:
41103209
NACRES:
NB.43

$54.70


Estimated to ship on24 April 2025


material

low-density polyethylene bottle
white polypropylene closure
wide-neck

packaging

pkg of 5 ea

manufacturer/tradename

Azlon WGW230P

capacity

250 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for solvent and water dispensing within the laboratory

Features and Benefits

Precision jet tip for fine control

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service