Skip to Content
Merck
All Photos(1)

Key Documents

DWKWGF534P

Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture

label, I.M.S, low-density polyethylene bottle, capacity 250 mL

Synonym(s):

WGF534P

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$57.10

$57.10


Estimated to ship on21 April 2025



Select a Size

Change View
5 EA
$57.10

About This Item

UNSPSC Code:
41103209
NACRES:
NB.43

$57.10


Estimated to ship on21 April 2025


material

low-density polyethylene bottle

feature

label I.M.S

packaging

pkg of 5 ea

manufacturer/tradename

Azlon WGF534P

capacity

250 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for IMS dispensing within the laboratory

Features and Benefits

Slim design for comfortable handling and ease of use

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service