Skip to Content
Merck
All Photos(2)

Key Documents

DWKBWH2000P

Azlon HDPE Wide Neck Round Bottles

screw cap, capacity 2000 mL

Synonym(s):

BWH2000P

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$130.00

$130.00


Check Cart for Availability


Select a Size

Change View
5 EA
$130.00

About This Item

UNSPSC Code:
41121803
NACRES:
NB.43

$130.00


Check Cart for Availability

material

screw cap
wide-neck

sterility

non-sterile

packaging

pkg of 5 ea

manufacturer/tradename

Azlon BWH2000P

capacity

2000 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for a wide range of liquid storage and sampling applications

Features and Benefits

Suitable for food contact applications

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service