Skip to Content
Merck
All Photos(2)

Key Documents

DWKBGS040

Azlon Polyamide Coated Steel Bottle Carrier

to hold, up to six 60mm diameter bottles

Synonym(s):

BGS040

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41000000
NACRES:
NB.43

feature

to hold up to six 60mm diameter bottles

packaging

pkg of 1 ea

manufacturer/tradename

Azlon BGS040

Looking for similar products? Visit Product Comparison Guide

Application

For safely carrying laboratory bottles

Features and Benefits

Good heat and chemical resistance

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service