Skip to Content
Merck
All Photos(2)

Key Documents

DWK292432883

Bottle Tag, For Gl 45 Duran® Media Bottles Bottle Tag

white silicone bottle

Synonym(s):

292432883

Sign Into View Organizational & Contract Pricing

Select a Size

20 EACH
$91.50

$91.50


Check Cart for Availability


Select a Size

Change View
20 EACH
$91.50

About This Item

UNSPSC Code:
41121800
NACRES:
NB.35

$91.50


Check Cart for Availability

material

white silicone bottle

packaging

pkg of 20 ea

manufacturer/tradename

DURAN® 292432883

Looking for similar products? Visit Product Comparison Guide

General description

The colourful GL 45 Bottle Tags offer an elegant solution to the marking of laboratory bottles. The Bottle Tags can be easily and securely attached around the neck of the YOUTILITY bottle, even with the screw cap in place. The Bottle Tags feature a graspable tab that makes them easy to remove with bare or gloved hands.

Application

easy customisation and unambiguous bottle identification.

Features and Benefits

for GL 45 bottles

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service