DWK213545505
Duran® Powder Funnel, Short Wide Stem
short, wide mouth stem(s)
Synonym(s):
213545505
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
material
short, wide mouth stem(s)
packaging
pkg of 1 ea
manufacturer/tradename
DURAN® 213545505
funnel diam.
160 mm
Looking for similar products? Visit Product Comparison Guide
Application
Transfering of powdered substances and granulated material,The spillage of substances is avoided, Transfering, Boro 3.3
Features and Benefits
With short and wide stem; Large stem diameter makes it suitable for transferring solids; Fired-on, highly durable white print
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service