Skip to Content
Merck
All Photos(2)

Key Documents

DWK213526105

Duran® Funnel Ribbed

Synonym(s):

213526105

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$198.00

$198.00


Check Cart for Availability


Select a Size

Change View
1 EACH
$198.00

About This Item

UNSPSC Code:
41121809
NACRES:
NB.43

$198.00


Check Cart for Availability

packaging

pkg of 1 ea

manufacturer/tradename

DURAN® 213526105

funnel diam.

200 mm

Looking for similar products? Visit Product Comparison Guide

Application

Decanting and filtration of liquids, The spillage of substances is avoided, Filtering, Boro 3.3

Features and Benefits

Due to its shape, it is suitable for liquid-solid filtration using round filter paper; Fired-on,

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service