DWK213116309
Duran® Crystallizing Dish
cylinder capacity 3500 mL
Synonym(s):
213116309
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
With Spout
packaging
pkg of 10 ea
manufacturer/tradename
DURAN® 213116309
cylinder capacity
3500 mL
Looking for similar products? Visit Product Comparison Guide
Application
heating and cooling baths, crystallization, disposal by evaporation and sand baths for microscale experiments
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service